2,6-Difluoro-3,5-dimethoxyaniline 97%
|
|
BD304761-100MG
BD304761-250MG
BLDPBD304761-100MGEA
8.4
EUR
BLDPBD304761-100MG
BLDPBD304761-250MG
2,6-Difluoro-3,5-dimethoxyaniline 97%
2,6-Difluoro-3,5-dimethoxyaniline
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₈H₉F₂NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD12973804 CAS Number: 651734-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...