(6-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%

BD305300-100MG BD305300-1G
BLDPBD305300-100MGEA 207 EUR
BLDPBD305300-100MG BLDPBD305300-1G
(6-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%
(6-(Difluoromethoxy)pyridin-3-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₆BF₂NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18261896
CAS Number: 1354290-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR