2-Cyclopropyl-1H-benzimidazole 97%

BD307457-1G BD307457-10G
BLDPBD307457-1GEA 332 EUR
BLDPBD307457-1G BLDPBD307457-10G
2-Cyclopropyl-1H-benzimidazole 97%
2-Cyclopropyl-1H-benzimidazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₀N₂
Storage Temperature: Refrigerator
MDL Number: MFCD01859172
CAS Number: 16405-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR