(R)-Piperidine-3-carboxylic acid amide 97%
|
|
BD307483-1G
BD307483-5G
BLDPBD307483-1GEA
66.2
EUR
BLDPBD307483-1G
BLDPBD307483-5G
(R)-Piperidine-3-carboxylic acid amide 97%
(R)-Piperidine-3-carboxylic acid amide
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₆H₁₂N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD17015910 CAS Number: 168749-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...