(S)-(+)-4-Phenyl-2-butanol 98%

BD307676-100MG BD307676-250MG
BLDPBD307676-100MGEA 35.7 EUR
BLDPBD307676-100MG BLDPBD307676-250MG
(S)-(+)-4-Phenyl-2-butanol 98%
(S)-(+)-4-Phenyl-2-butanol

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD03093082
CAS Number: 22148-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR