4-Amino-2-chloronicotinic acid 97%

BD314062-100MG BD314062-10G
BLDPBD314062-100MGEA 125 EUR
BLDPBD314062-100MG BLDPBD314062-10G
4-Amino-2-chloronicotinic acid 97%
4-Amino-2-chloronicotinic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13188881
CAS Number: 1018678-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR