5-Bromo-2-iodobenzyl alcohol 97%

BD315438-10G BD315438-25G
BLDPBD315438-10GEA 550 EUR
BLDPBD315438-10G BLDPBD315438-25G
5-Bromo-2-iodobenzyl alcohol 97%
5-Bromo-2-iodobenzyl alcohol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₆BrIO
Storage Temperature: Ambient
MDL Number: MFCD09745850
CAS Number: 199786-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR