4-Bromo-5-chloropyridin-2-amine 95%
|
|
BD316424-100MG
BD316424-1G
BLDPBD316424-100MGEA
16.8
EUR
BLDPBD316424-100MG
BLDPBD316424-1G
4-Bromo-5-chloropyridin-2-amine 95%
4-Bromo-5-chloropyridin-2-amine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₅H₄BrClN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD15143360 CAS Number: 1187449-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...