Levulinic acid 97%

BD31667-500G BD31667-100G
BLDPBD31667-500GEA 30.5 EUR
BLDPBD31667-500G BLDPBD31667-100G
Levulinic acid 97%
Levulinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00002796
CAS Number: 123-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR