4-Bromo-3,5-difluorophenol 98%

BD31720-10G BD31720-1G
BLDPBD31720-10GEA 156 EUR
BLDPBD31720-10G BLDPBD31720-1G
4-Bromo-3,5-difluorophenol 98%
4-Bromo-3,5-difluorophenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₃BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD08062385
CAS Number: 130191-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR