4-(1-Methyl-4-piperidyl)phenylboronic Acid Pinacol Ester 98%
|
|
BD317284-5G
BD317284-25G
BLDPBD317284-5GEA
341
EUR
BLDPBD317284-5G
BLDPBD317284-25G
4-(1-Methyl-4-piperidyl)phenylboronic Acid Pinacol Ester 98%
4-(1-Methyl-4-piperidyl)phenylboronic Acid Pinacol Ester
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₈H₂₈BNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18383496 CAS Number: 1247000-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...