Methyl-3-cyanobenzoate 97%
|
|
BD31809-100G
BD31809-25G
BLDPBD31809-100GEA
111
EUR
BLDPBD31809-100G
BLDPBD31809-25G
Methyl-3-cyanobenzoate 97%
Methyl 3-cyanobenzoate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₇NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00220156 CAS Number: 13531-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...