2,4-Dibromo-5-fluoropyrimidine 95%

BD318311-5G BD318311-100G
BLDPBD318311-5GEA 39.9 EUR
BLDPBD318311-5G BLDPBD318311-100G
2,4-Dibromo-5-fluoropyrimidine 95%
2,4-Dibromo-5-fluoropyrimidine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₄HBr₂FN₂
Storage Temperature: Refrigerator
MDL Number: MFCD21647860
CAS Number: 1372096-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR