Z-Aib-OH 97%

BD32020-5G BD32020-10G
BLDPBD32020-5GEA 7.4 EUR
BLDPBD32020-5G BLDPBD32020-10G
Z-Aib-OH 97%
Z-Aib-OH

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₅NO₄
MW: 237.255 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00004190
CAS Number: 15030-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR