Boc-3-nitro-D-phenylalanine 95%
|
|
BD32117-1G
BD32117-5G
BLDPBD32117-1GEA
53.6
EUR
BLDPBD32117-1G
BLDPBD32117-5G
Boc-3-nitro-D-phenylalanine 95%
Boc-3-nitro-D-phenylalanine
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₄H₁₈N₂O₆ Storage Temperature: Refrigerator |
MDL Number:
MFCD01317702 CAS Number: 158741-21-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...