6-Bromopiperonal 98%

BD32130-500G BD32130-1G
BLDPBD32130-500GEA 1200 EUR
BLDPBD32130-500G BLDPBD32130-1G
6-Bromopiperonal 98%
6-Bromopiperonal

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₅BrO₃
Storage Temperature: Ambient
MDL Number: MFCD00022952
CAS Number: 15930-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR