2-Bromo-3-fluorophenol 98%
|
|
BD3219-25G
BD3219-10G
BLDPBD3219-25GEA
109
EUR
BLDPBD3219-25G
BLDPBD3219-10G
2-Bromo-3-fluorophenol 98%
2-Bromo-3-fluorophenol
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₄BrFO Storage Temperature: Ambient |
MDL Number:
MFCD04037220 CAS Number: 443-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...