Fmoc-D-Glu-OH 95%

BD322095-5G BD322095-10G
BLDPBD322095-5GEA 14.7 EUR
BLDPBD322095-5G BLDPBD322095-10G
Fmoc-D-Glu-OH 95%
Fmoc-D-Glu-OH

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₂₀H₁₉NO₆
Storage Temperature: Ambient
MDL Number: MFCD09037362
CAS Number: 104091-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR