tert-Butyl (2-oxopropyl)carbamate 97%

BD32249-100G BD32249-250MG
BLDPBD32249-100GEA 928 EUR
BLDPBD32249-100G BLDPBD32249-250MG
tert-Butyl (2-oxopropyl)carbamate 97%
tert-Butyl (2-oxopropyl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₅NO₃
MW: 173.2096 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10698678
CAS Number: 170384-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR