2-Bromo-6-nitrophenol 98%

BD3226-5G BD3226-25G
BLDPBD3226-5GEA 7.4 EUR
BLDPBD3226-5G BLDPBD3226-25G
2-Bromo-6-nitrophenol 98%
2-Bromo-6-nitrophenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₄BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD02683216
CAS Number: 13073-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR