3-Fluoro-5-formylbenzeneboronic acid pinacol ester 97%
|
|
BD324898-1G
BD324898-100MG
BLDPBD324898-1GEA
185
EUR
BLDPBD324898-1G
BLDPBD324898-100MG
3-Fluoro-5-formylbenzeneboronic acid pinacol ester 97%
3-Fluoro-5-formylbenzeneboronic acid pinacol ester
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₃H₁₆BFO₃ MW: 250.07 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16996301 CAS Number: 1112209-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...