a-Amino-4-methoxybenzeneacetic acid 95%

BD32495-1G BD32495-250MG
BLDPBD32495-1GEA 52.5 EUR
BLDPBD32495-1G BLDPBD32495-250MG
a-Amino-4-methoxybenzeneacetic acid 95%
a-Amino-4-methoxybenzeneacetic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049329
CAS Number: 19789-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR