5-Chloro-2,1,3-benzothiadiazol-4-ylamine 98%

BD3250-500G BD3250-1G
BLDPBD3250-500GEA 1060 EUR
BLDPBD3250-500G BLDPBD3250-1G
5-Chloro-2,1,3-benzothiadiazol-4-ylamine 98%
5-Chloro-2,1,3-benzothiadiazol-4-ylamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₄ClN₃S
Storage Temperature: Ambient
MDL Number: MFCD00465908
CAS Number: 30536-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR