3-(4-Nitrophenyl)-1,2,4-oxadiazole 98%
|
|
BD326788-5G
BD326788-1G
BLDPBD326788-5GEA
335
EUR
BLDPBD326788-5G
BLDPBD326788-1G
3-(4-Nitrophenyl)-1,2,4-oxadiazole 98%
3-(4-Nitrophenyl)-1,2,4-oxadiazole
Storage condition: Sealed in dry, 2-8°C
Formula:
C₈H₅N₃O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00274808 CAS Number: 16013-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...