5-Bromo-2-hydroxy-3-nitro-4-picoline 98%
|
|
BD32744-250MG
BD32744-25G
BLDPBD32744-250MGEA
7.4
EUR
BLDPBD32744-250MG
BLDPBD32744-25G
5-Bromo-2-hydroxy-3-nitro-4-picoline 98%
5-Bromo-2-hydroxy-3-nitro-4-picoline
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₅BrN₂O₃ MW: 233.0200 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659520 CAS Number: 228410-90-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...