8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 97%
|
|
BD329639-1G
BD329639-10G
BLDPBD329639-1GEA
88.2
EUR
BLDPBD329639-1G
BLDPBD329639-10G
8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one 97%
8-Fluoro-4,5-dihydro-1H-azepino[5,4,3-cd]indol-6(3H)-one
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₁H₉FN₂O Storage Temperature: Ambient |
MDL Number:
MFCD23381312 CAS Number: 1408282-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...