2,4-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 95%
|
|
BD329681-100MG
BD329681-250MG
BLDPBD329681-100MGEA
41
EUR
BLDPBD329681-100MG
BLDPBD329681-250MG
2,4-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 95%
2,4-Difluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₁₂H₁₆BF₂NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD28291811 CAS Number: 1415568-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...