9,10-Di(1-naphthyl)anthracene 97%

BD32974-25G BD32974-250MG
BLDPBD32974-25GEA 260 EUR
BLDPBD32974-25G BLDPBD32974-250MG
9,10-Di(1-naphthyl)anthracene 97%
9,10-Di(1-naphthyl)anthracene

Storage condition: Sealed in dry, Room Temperature

Formula: C₃₄H₂₂
Storage Temperature: Ambient
MDL Number: MFCD04222155
CAS Number: 26979-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR