3,4-Thiophenedicarboxylic acid 98%

BD3298-25G BD3298-10G
BLDPBD3298-25GEA 165 EUR
BLDPBD3298-25G BLDPBD3298-10G
3,4-Thiophenedicarboxylic acid 98%
3,4-Thiophenedicarboxylic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₄O₄S
MW: 172.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067079
CAS Number: 4282-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR