N-(2-Cyanophenyl)picolinamide 95%

BD331215-1G BD331215-250MG
BLDPBD331215-1GEA 67.2 EUR
BLDPBD331215-1G BLDPBD331215-250MG
N-(2-Cyanophenyl)picolinamide 95%
N-(2-Cyanophenyl)picolinamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₉N₃O
Storage Temperature: Ambient
MDL Number: MFCD02128063
CAS Number: 304650-02-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR