1-Amino-3,3-dimethyl-butan-2-one hydrochloride 98%
|
|
BD33242-100G
BD33242-10G
BLDPBD33242-100GEA
612
EUR
BLDPBD33242-100G
BLDPBD33242-10G
1-Amino-3,3-dimethyl-butan-2-one hydrochloride 98%
1-Amino-3,3-dimethyl-butan-2-one hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₁₃NO·ClH Storage Temperature: Ambient |
MDL Number:
MFCD06798286 CAS Number: 33119-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...