2-Amino-3,5-dibromo-4-methylpyridine 97%
|
|
BD33301-500G
BD33301-10G
BLDPBD33301-500GEA
348
EUR
BLDPBD33301-500G
BLDPBD33301-10G
2-Amino-3,5-dibromo-4-methylpyridine 97%
2-Amino-3,5-dibromo-4-methylpyridine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₅H₄Br₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD01646065 CAS Number: 3430-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...