2-Bromo-5-fluoro-6-methylpyridine 98%

BD3331-1G BD3331-250MG
BLDPBD3331-1GEA 10.5 EUR
BLDPBD3331-1G BLDPBD3331-250MG
2-Bromo-5-fluoro-6-methylpyridine 98%
2-Bromo-5-fluoro-6-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₅BrFN
MW: 190.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095064
CAS Number: 374633-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR