2-Chloro-5-fluoropyridin-4-ol 97%

BD333347-25G BD333347-1G
BLDPBD333347-25GEA 969 EUR
BLDPBD333347-25G BLDPBD333347-1G
2-Chloro-5-fluoropyridin-4-ol 97%
2-Chloro-5-fluoropyridin-4-ol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₃ClFNO
Storage Temperature: Refrigerator
MDL Number: MFCD13189882
CAS Number: 1196153-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR