5-Methoxy-3-methylpicolinonitrile 98%

BD334043-100MG BD334043-1G
BLDPBD334043-100MGEA 119 EUR
BLDPBD334043-100MG BLDPBD334043-1G
5-Methoxy-3-methylpicolinonitrile 98%
5-Methoxy-3-methylpicolinonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD18255402
CAS Number: 1256792-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR