4,5-Dihydroxy-m-anisic acid 97%

BD33516-5G BD33516-25G
BLDPBD33516-5GEA 208 EUR
BLDPBD33516-5G BLDPBD33516-25G
4,5-Dihydroxy-m-anisic acid 97%
4,5-Dihydroxy-m-anisic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₈O₅
Storage Temperature: Ambient
MDL Number: MFCD00016518
CAS Number: 3934-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR