α'-Bromo-α,α,α-trifluoro-o-xylene 98%
|
|
BD33526-25G
BD33526-10G
BLDPBD33526-25GEA
22.1
EUR
BLDPBD33526-25G
BLDPBD33526-10G
α'-Bromo-α,α,α-trifluoro-o-xylene 98%
α'-Bromo-α,α,α-trifluoro-o-xylene
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₈H₆BrF₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00013558 CAS Number: 395-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...