2-Chloro-3-fluoro-5-methylpyridine 98%
|
|
BD3354-500G
BD3354-5G
BLDPBD3354-500GEA
2120
EUR
BLDPBD3354-500G
BLDPBD3354-5G
2-Chloro-3-fluoro-5-methylpyridine 98%
2-Chloro-3-fluoro-5-methylpyridine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₅ClFN Storage Temperature: Refrigerator |
MDL Number:
MFCD06658238 CAS Number: 34552-15-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...