2-Chloro-4-methyl-3-nitropyridine 98%

BD3361-500G BD3361-5G
BLDPBD3361-500GEA 455 EUR
BLDPBD3361-500G BLDPBD3361-5G
2-Chloro-4-methyl-3-nitropyridine 98%
2-Chloro-4-methyl-3-nitropyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00012347
CAS Number: 23056-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR