DL-Valine 98%

BD33893-500G BD33893-100G
BLDPBD33893-500GEA 93.5 EUR
BLDPBD33893-500G BLDPBD33893-100G
DL-Valine 98%
DL-Valine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00004267
CAS Number: 516-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR