Atorvastatin calcium salt trihydrate 95%
|
|
BD340761-5G
BD340761-100G
BLDPBD340761-5GEA
31.5
EUR
BLDPBD340761-5G
BLDPBD340761-100G
Atorvastatin calcium salt trihydrate 95%
Atorvastatin calcium salt trihydrate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆₆H₇₄CaF₂N₄O₁₃ Storage Temperature: Ambient |
MDL Number:
MFCD09752074 CAS Number: 344423-98-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...