2-Chloro-3-fluoroisonicotinic acid 97%

BD34406-1G BD34406-500G
BLDPBD34406-1GEA 6.3 EUR
BLDPBD34406-1G BLDPBD34406-500G
2-Chloro-3-fluoroisonicotinic acid 97%
2-Chloro-3-fluoroisonicotinic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD04038325
CAS Number: 628691-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR