3-Bromo-2-methox-6-methylypyridine 98%

BD34724-100G BD34724-250MG
BLDPBD34724-100GEA 1700 EUR
BLDPBD34724-100G BLDPBD34724-250MG
3-Bromo-2-methox-6-methylypyridine 98%
3-Bromo-2-methox-6-methylypyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD04112515
CAS Number: 717843-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR