Ethyl-8-chloro-4-hydroxyquinoline-3-carboxylate 97%

BD34792-5G BD34792-1G
BLDPBD34792-5GEA 109 EUR
BLDPBD34792-5G BLDPBD34792-1G
Ethyl-8-chloro-4-hydroxyquinoline-3-carboxylate 97%
Ethyl 8-chloro-4-hydroxyquinoline-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₀ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00024002
CAS Number: 73987-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR