2-Fluoro-6-methylpyridine 97%

BD3493-10G BD3493-25G
BLDPBD3493-10GEA 13.7 EUR
BLDPBD3493-10G BLDPBD3493-25G
2-Fluoro-6-methylpyridine 97%
2-Fluoro-6-methylpyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₆FN
Storage Temperature: Ambient
MDL Number: MFCD00041226
CAS Number: 407-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR