Methyl 2-Hydroxycyclopentylcarboxylate 98%
|
|
BD35463-25G
BD35463-1G
BLDPBD35463-25GEA
508
EUR
BLDPBD35463-25G
BLDPBD35463-1G
Methyl 2-Hydroxycyclopentylcarboxylate 98%
Methyl 2-Hydroxycyclopentylcarboxylate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₇H₁₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD04972392 CAS Number: 90085-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...