3,4,5-Trifluoro-L-phenylalanine 97%

BD36085-100MG BD36085-250MG
BLDPBD36085-100MGEA 69.3 EUR
BLDPBD36085-100MG BLDPBD36085-250MG
3,4,5-Trifluoro-L-phenylalanine 97%
3,4,5-Trifluoro-L-phenylalanine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD03428422
CAS Number: 646066-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR