(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97%
|
|
BD3641-25G
BD3641-5G
BLDPBD3641-25GEA
448
EUR
BLDPBD3641-25G
BLDPBD3641-5G
(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97%
(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₆H₇NO Storage Temperature: Ambient |
MDL Number:
MFCD00211275 CAS Number: 130931-83-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...