Benzyl 2-(2-benzyl-3-mercaptopropanamido)acetate 95+%

BD377035-50MG BD377035-10MG
BLDPBD377035-50MGEA 1300 EUR
BLDPBD377035-50MG BLDPBD377035-10MG
Benzyl 2-(2-benzyl-3-mercaptopropanamido)acetate 95+%
Benzyl 2-(2-benzyl-3-mercaptopropanamido)acetate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formula: C₁₉H₂₁NO₃S
Storage Temperature: Freezer
MDL Number: MFCD28009480
CAS Number: 81110-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR