2-Bromo-4-fluorobenzonitrile 97%

BD4026-500G BD4026-5G
BLDPBD4026-500GEA 439 EUR
BLDPBD4026-500G BLDPBD4026-5G
2-Bromo-4-fluorobenzonitrile 97%
2-Bromo-4-fluorobenzonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₃BrFN
Storage Temperature: Ambient
MDL Number: MFCD00672924
CAS Number: 36282-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR